Allsigns International™ PVC Hazardous Label


Overview
Brand: Allsigns International™

Manufacturer Part Number: HZ7

Hazard label diamonds PVC self adhesive 100mm x100mm Corrosive Allsigns, 1/Cs, MA: TBD

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12365778

Quantity Price
1 £ 93.75 / 1 roll
4 £ 75.0 / 1 roll
Estimated Shipment
06-02-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Ideal for Specific Uses Wherever Hazardous Materials are in Use
Alphanumeric Character Corrosive
Width (Metric) 100mm
Quantity 1Each
Type Hazardous Label
Adhesive Type Self
Message Corrosive
Resistant To Chemicals
Color Black Markings on White Background
Material PVC
Dimensions (L x W) 100 x 100mm
Height (Metric) 100mm
Shape Diamond
Length (Metric) 100mm

Sold in single rolls, these hazard diamonds are ideal for specific uses wherever hazardous materials are in use. In self adhesive vinyl.

Documentation