Allsigns International™ PVC Hazardous Label


Overview
Brand: Allsigns International™

Manufacturer Part Number: RO6

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12385788

Quantity Price
1 £ 87.78 / Pack of 250
4 £ 79.0 / Pack of 250
Estimated Shipment
15-03-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Invaluable for Marking Hazards on Containers, Instruments and Equipments
Width (Metric) 50mm
Alphanumeric Character Beware
Quantity 250Each
Type Hazardous Label
Adhesive Type Self
Material PVC
Color Black with Yellow on White Background
Message Beware Caution
Resistant To Chemicals
Height (Metric) 50mm
Dimensions (L x W) 50 x 50mm
Length (Metric) 50mm
Shape Square

Sold in single rolls, these hazard diamonds are ideal for specific uses wherever hazardous materials are in use. In self adhesive vinyl.

Documentation