missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ PVC Hazardous Label Roll

Mark hazardous material warning messages on containers, instruments and equipment when hazardous materials are in use. Each Allsigns International™ PVC Hazardous Label Roll features a distinct warning message on a roll of self-adhesive vinyl “diamond” labels.

Brand:  Allsigns International™ DA25

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12375768

  • POA / Each


Estimated Shipment
10-08-2017
Add to basket

Specifications

Description

  • Sold in single rolls
  • Self-adhesive vinyl
  • Perforated for ease of removal
Description & Specifications

Specifications

Material: Perforated vinyl
Poison 6
Vinyl
102mm
102 mm
Chemicals
Documentation