Allsigns International™ PVC Hazardous Label


Overview
Brand: Allsigns International™

Manufacturer Part Number: BUR-520-Q

Hazard label diamonds PVC self adhesive 100mm x100mm Radioactive Allsigns, 1/Cs, MA: TBD

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12345778

Quantity Price
1 £ 94.06 / 1 roll
4 £ 75.25 / 1 roll
Estimated Shipment
10-03-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Ideal for Specific Uses Wherever Hazardous Materials are in Use
Alphanumeric Character Radioactive 7
Width (Metric) 100mm
Adhesive Type Self
Quantity 1Each
Type Hazardous Label
Resistant To Chemicals
Color Yellow on White With Black Markings
Material PVC
Message Radioactive 7
Dimensions (L x W) 100 x 100mm
Shape Diamond
Length (Metric) 100mm

Sold in single rolls, these hazard diamonds are ideal for specific uses wherever hazardous materials are in use. In self adhesive vinyl.

Documentation