Allsigns International™ PVC Hazardous Label


Overview
Brand: Allsigns International™

Manufacturer Part Number: R02

S/LABELSON ROLL TOXIC

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12365788

Quantity Price
1 £ 85.61 / 1 roll
Estimated Shipment
07-02-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Ideal for Specific Uses Wherever Hazardous Materials are in Use
Alphanumeric Character Toxic Hazard
Width (Metric) 50mm
Adhesive Type Self
Quantity 250Each
Type Hazardous Label
Resistant To Chemicals
Color Black with Yellow on White Background
Material PVC
Message Toxic Hazard
Dimensions (L x W) 50 x 50mm
Height (Metric) 50mm
Shape Square
Length (Metric) 50mm

Sold in single rolls, these hazard diamonds are ideal for specific uses wherever hazardous materials are in use. In self adhesive vinyl.

Documentation