missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ PVC Hazard Label Safety Floor Marking Tape

Brand:  Allsigns International™ FMT7

UNSPSC : 46161507

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12349727

  • £33.24 / Each


Estimated Shipment
12-07-2017
Add to basket

Specifications Description & Specifications

Specifications

50mm x 33m
Blue
Safety Floor Marking Tape
Self-adhesive
1Each
PVC
Highlighting hazards/restricted areas
Documentation

Technical Documents