missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ PVC Hazard Label Safety Floor Marking Tape

Brand:  Allsigns International™ FMT6

Code : 04

Additional Details : Weight : 0.32524kg

 View more versions of this product

Product Code. 12339727

  • £33.62 / Each


In Stock 1
Add to basket

You may only order up to the available stock. For more details call customer services on 01509 555 500

Specifications

Specifications

50mm x 33m
Green
Safety Floor Marking Tape
Self-adhesive
1Each
PVC
Highlighting hazards/restricted areas
Videos
Documentation

Technical Documents