missing translation for 'onlineSavingsMsg'
Learn More

Purvalanol A, Tocris Bioscience™

Cdk inhibitor; potently inhibits cdk1, cdk2 and cdk5

Brand:  Tocris (Bio-Techne) 158050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13449713

  • £840.84 / 50mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cyclin-dependent kinase (cdk) inhibitor (reported IC50 values are 4, 4 - 70, 75 - 240 and 100 nM for cdk1, cdk2, cdk5 and cdk 7, respectively). Reversibly arrests synchronised cells in G1 and G2, and inhibits cell proliferation and cell death.
Specifications

Chemical Identifiers

212844-53-6
388.9
purvalanol a, purv, 6-3-chloro anilino-2-isopropyl-2-hydroxyethylamino-9-isopropylpurine, chembl23327, 2-1r-isopropyl-2-hydroxyethylamino-6-3-chloroanilino-9-isopropylpurine, 2r-2-6-3-chlorophenyl amino-9-propan-2-yl-9h-purin-2-yl amino-3-methylbutan-1-ol, 2r-2-6-3-chloroanilino-9-propan-2-ylpurin-2-yl amino-3-methylbutan-1-ol, 2-6-3-chlorophenyl amino-9-isopropyl-9h-purin-2-yl amino-3-methylbutan-1-ol, tocris-1580, purvalanol a ng 60
CHEBI:47600
CC(C)C(CO)NC1=NC2=C(C(=N1)NC3=CC(=CC=C3)Cl)N=CN2C(C)C
C19H25ClN6O
PMXCMJLOPOFPBT-HNNXBMFYSA-N
456214
(2R)-2-[[6-(3-chloroanilino)-9-propan-2-ylpurin-2-yl]amino]-3-methylbutan-1-ol

Specifications

50mg
212844-53-6
purvalanol a, purv, 6-3-chloro anilino-2-isopropyl-2-hydroxyethylamino-9-isopropylpurine, chembl23327, 2-1r-isopropyl-2-hydroxyethylamino-6-3-chloroanilino-9-isopropylpurine, 2r-2-6-3-chlorophenyl amino-9-propan-2-yl-9h-purin-2-yl amino-3-methylbutan-1-ol, 2r-2-6-3-chloroanilino-9-propan-2-ylpurin-2-yl amino-3-methylbutan-1-ol, 2-6-3-chlorophenyl amino-9-isopropyl-9h-purin-2-yl amino-3-methylbutan-1-ol, tocris-1580, purvalanol a ng 60
CC(C)C(CO)NC1=NC2=C(C(=N1)NC3=CC(=CC=C3)Cl)N=CN2C(C)C
388.9
CHEBI:47600
>98%
Purvalanol A
C19H25ClN6O
PMXCMJLOPOFPBT-HNNXBMFYSA-N
(2R)-2-[[6-(3-chloroanilino)-9-propan-2-ylpurin-2-yl]amino]-3-methylbutan-1-ol
456214
388.9
Videos
Documents