missing translation for 'onlineSavingsMsg'
Learn More

Purvalanol A, Tocris Bioscience™

Cdk inhibitor; potently inhibits cdk1, cdk2 and cdk5

Brand:  RD Systems (Bio-Techne) 1580/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13439713

  • £196.68 / 10mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cyclin-dependent kinase (cdk) inhibitor (reported IC50 values are 4, 4 - 70, 75 - 240 and 100 nM for cdk1, cdk2, cdk5 and cdk 7, respectively). Reversibly arrests synchronised cells in G1 and G2, and inhibits cell proliferation and cell death.
Specifications

Chemical Identifiers

212844-53-6
388.9
purvalanol a, purv, 6-3-chloro anilino-2-isopropyl-2-hydroxyethylamino-9-isopropylpurine, chembl23327, 2-1r-isopropyl-2-hydroxyethylamino-6-3-chloroanilino-9-isopropylpurine, 2r-2-6-3-chlorophenyl amino-9-propan-2-yl-9h-purin-2-yl amino-3-methylbutan-1-ol, 2r-2-6-3-chloroanilino-9-propan-2-ylpurin-2-yl amino-3-methylbutan-1-ol, 2-6-3-chlorophenyl amino-9-isopropyl-9h-purin-2-yl amino-3-methylbutan-1-ol, tocris-1580, purvalanol a ng 60
CHEBI:47600
CC(C)C(CO)NC1=NC2=C(C(=N1)NC3=CC(=CC=C3)Cl)N=CN2C(C)C
C19H25ClN6O
PMXCMJLOPOFPBT-HNNXBMFYSA-N
456214
(2R)-2-[[6-(3-chloroanilino)-9-propan-2-ylpurin-2-yl]amino]-3-methylbutan-1-ol

Specifications

Purvalanol A
212844-53-6
purvalanol a, purv, 6-3-chloro anilino-2-isopropyl-2-hydroxyethylamino-9-isopropylpurine, chembl23327, 2-1r-isopropyl-2-hydroxyethylamino-6-3-chloroanilino-9-isopropylpurine, 2r-2-6-3-chlorophenyl amino-9-propan-2-yl-9h-purin-2-yl amino-3-methylbutan-1-ol, 2r-2-6-3-chloroanilino-9-propan-2-ylpurin-2-yl amino-3-methylbutan-1-ol, 2-6-3-chlorophenyl amino-9-isopropyl-9h-purin-2-yl amino-3-methylbutan-1-ol, tocris-1580, purvalanol a ng 60
CC(C)C(CO)NC1=NC2=C(C(=N1)NC3=CC(=CC=C3)Cl)N=CN2C(C)C
388.9
CHEBI:47600
>98%
10mg
C19H25ClN6O
PMXCMJLOPOFPBT-HNNXBMFYSA-N
(2R)-2-[[6-(3-chloroanilino)-9-propan-2-ylpurin-2-yl]amino]-3-methylbutan-1-ol
456214
388.9
Videos