missing translation for 'onlineSavingsMsg'
Learn More

Purpurin, Thermo Scientific Chemicals

Catalog Number p-188876
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
81-54-9
C14H8O5
256.2
MFCD00001203
BBNQQADTFFCFGB-UHFFFAOYSA-N
purpurin, 1,2,4-trihydroxyanthraquinone, purpurine, verantin, hydroxylizaric acid, smoke brown g, 1,2,4-trihydroxy-9,10-anthracenedione, 1,2,4-trihydroxy-9,10-anthraquinone, 1,2,4-trihydroxyanthrachinon, 9,10-anthracenedione, 1,2,4-trihydroxy
6683
CHEBI:8645
1,2,4-trihydroxyanthracene-9,10-dione
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3O)O)O
This item is not returnable. View return policy
81-54-9
C14H8O5
256.2
MFCD00001203
BBNQQADTFFCFGB-UHFFFAOYSA-N
purpurin, 1,2,4-trihydroxyanthraquinone, purpurine, verantin, hydroxylizaric acid, smoke brown g, 1,2,4-trihydroxy-9,10-anthracenedione, 1,2,4-trihydroxy-9,10-anthraquinone, 1,2,4-trihydroxyanthrachinon, 9,10-anthracenedione, 1,2,4-trihydroxy
6683
CHEBI:8645
1,2,4-trihydroxyanthracene-9,10-dione
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
Melting Point 253°C to 256°C
Color Brown to Brown-Red
Flash Point 113°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Beilstein 08, 509
Merck Index 15, 8056
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in hot water,freely soluble in alcohol and ether,soluble in benzene,toluene; xylene,boiling alum,solution
Formula Weight 256.2
Physical Form Powder
Chemical Name or Material Purpurin
Show More Show Less
compliance-icons
Product Identifier
  • Purpurin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'