Learn More
Invitrogen™ Puromycin N-acetyltransferase Recombinant Rabbit Monoclonal Antibody (1H12L8)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 702389
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Puromycin N-acetyl transferase functions to detox the cell of puromycin and plays a role in antibiotic resistance.
Specifications
| Puromycin N-acetyltransferase | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
| Western Blot | |
| 1H12L8 | |
| Unconjugated | |
| Puromycin N-acetyl-transferase | |
| Protein corresponding to Streptomyces alboniger Pac (aa 1-199). | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Responsibly packaged |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.