missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Puromycin Dihydrochloride,, MP Biomedicals
Click to view available options
Quantity:
10 mg
25 mg
100 mg
250 mg
500 mg
1 g
Specifications
Type | Puromycin Dihydrochloride |
Physical Form | Powder |
Extinction Coefficient | 20.3 (275nm, 0.1N NaOH), 19.5 (268nm, 0.1 NHCl) (Lit.) |
IUPAC Name | (2S)-2-amino-N-[(2S,3S,4R,5R)-5-[6-(dimethylamino)-9H-purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide dihydrochloride |
Formula Weight | 544.4 |
Color | White |
Quantity | 10 mg |
Chemical Name or Material | Puromycin Dihydrochloride, from Streptomyces albo-niger |
Safety and Handling
Product Identifier
- Puromycin Dihydrochloride
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Puromycin Dihydrochloride
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'