missing translation for 'onlineSavingsMsg'
Learn More

Pure Deionised Water, Waternation

Product Code. 17878030
Click to view available options
Quantity:
10 x 100 mL
10 x 500 mL
6 x 1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

EP grade Pure Deionised Water. Complies with the requirements of ASTM D1193-06, ISO 3696 Type 2A and 3A.

  • Ready to use
  • No chemical additives
  • Batch traceability
  • Certificate of conformity and analysis
  • Reduce expensive test fails due to impurities
  • High performance
  • High accuracy
  • Suppress water-forming side reactions
  • Provide consistent performance across many sample types
The MLG range, a comprehensive suite of five high grade pure waters, in assorted sizes, ranging from simple deionised to molecular biology grades, catering to the majority of pure water lab uses.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

pH 7
Conductivity 1 μS/cm
Grade EP
Packaging PET Plastic Bottle
Certificate of Analysis Yes
Quantity 10 x 100 mL
For Use With (Application) Glassware Washing and Rinsing
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'