missing translation for 'onlineSavingsMsg'
Learn More

Retsch™ Pure 70mm Diameter Stainless Steel Ring Sieve For Ultra Centrifugal Mill ZM 200

Brand:  Retsch™ 03.647.0292

Code : 50

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 10650974

  • £171.00 / Each


Estimated Shipment
29-03-2018
Add to basket

Specifications

Specifications

1Each
1mm
Stainless Steel
70mm
Ring Sieves
Ultra Centrifugal Mill ZM 200
Videos
Documentation