missing translation for 'onlineSavingsMsg'
Learn More

Whatman™ Puradisc™ Syringeless Filters

Brand:  Whatman™ 6799-2504

Code : A0

Additional Details : Weight : 1.35000kg

Product Code. 11989227

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

Puradisc Syringe Filters combine premium quality and economy. They are well suited for quick, efficient filtration of samples up to 100mL volume. Puradisc filters are produced from pigment-free polypropylene or polycarbonate with standard inlet (female luer lock) and outlet (male luer) connections, unless otherwise stated. Options include a sterile, medical-grade blister pack for critical applications and a special tube tip outlet that allows the sample to be accurately dispensed into a micro-vial, eliminating air lock.
  • Standard inlet and outlet luer connectors.
  • Optional sterile, medical-grade blister pack.
  • Tube-tip format (optional) for accurate dispensing into a micro-vial.
  • Choice of membrane or glass microfibre filter media.
  • Choice of filter sizes (4mm, 13mm, or 30mm) to minimise sample loss.
  • Sterile option for critical applications.
Specifications

Specifications

Puradisc Syringeless Filters
HPLC aqueous sample preparation, Biological sample preparation, Buffer solutions, Salt solutions, Tissue culture media, Irrigation solutions, Sterile isolation
.
0.45μm
500Each
No
25mm
Polypropylene
Nylon
4.2cm2
≤100mL
Videos