Learn More
Cytiva Whatman™ Puradisc PVDF Syringe Filters: Nonsterile
Designed for high tensile strength, high solvent resistance, and low protein binding. Cytiva Whatman™ Puradisc PVDF Syringe Filters: Nonsterile are well-suited for biomedical filtration, sterilization filtration, and HPLC sample preparation.
Brand: Cytiva 6749-2504
Code : A0
Additional Details : Weight : 2.65000kg
Description
- Manufactured from Polyvinylidene Difluoride (PVDF)
- Autoclavable
- Nonsterile
- Includes Female Luer Lock inlet and Male Luer outlet
Specifications
Puradisc 25 mm Polyvinylidene Difluoride Syringe Filter, 0.45 μm (1000 pcs) | |
PVDF | |
Non-sterile | |
HPLC aqueous sample preparation, Biological sample preparation, Buffer solutions, Salt solutions, Tissue culture media, Irrigation solutions, Sterile isolation . |
|
Female luer lock | |
1000Each | |
1000 Pieces | |
Puradisc Syringeless Filter | |
22.9 x 28.4 mm |
PVDF | |
0.45 μm | |
4.2 cm2 | |
Pigment Free Polypropylene (PP) | |
Male luer | |
5.2 bar (75 psi) | |
131°C | |
Up to 100 mL | |
25 mm |