missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ PTPMT1 Recombinant Human Protein, N-His Tag

Expressed from DNA sequence encoding human PTPMT1 isoform 1 (Lys 28-Thr 201) fused to polyhistidine tag at N-terminus

Brand:  Sino Biological™ 12902H07E5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15236085

  • £83.81 / 5µg


Estimated Shipment
10-07-2018
Add to basket

Description

Description

  • Formulation: PBS, 10% Glycerol, 1mM DTT, pH 7.4
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
114971
≥90% by SDS-PAGE
Human
Protein Tyrosine Phosphatase, Mitochondrial 1
Assay Standard
E.coli
His Tag
Lyophilized
21.7kd
5μg
Full Length, Recombinant
PTPMT1
Other Proteins
PTPMT1 Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.