missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ PTH Recombinant Human Protein, N-GST Tag

Expressed from DNA sequence encoding human PTH (Ser 32-Phe 65) fused to GST tag at N-terminus

Brand:  Sino Biological™ 13192H09E5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15266235

  • £55.70 / 5µg


Estimated Shipment
10-07-2018
Add to basket

Description

Description

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
5741
≥90% by SDS-PAGE
Human
<1 EU/μg
PTH
Hormones and Receptors
PTH Recombinant Human Protein
Lyophilized
31kd
5μg
Truncated, Recombinant
Parathyroid Hormone
Assay Standard
E.coli
GST Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.