missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ PTGS2 (COX-2) Recombinant Human Protein

Expressed from DNA sequence encoding human PTGS2 (NP_000954.1) (Met 1-Leu 604) fused to a polyhistidine tag at the C-terminus

Brand:  Sino Biological™ 12036H08B5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15440574

  • £83.68 / 5µg


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: 50mM Tris, 100mM NaCl, 0.5mM PMSF, 10% glycerol, pH 8.0
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
5743
≥90% by SDS-PAGE
Human
<1 EU/μg
PTGS2
Other proteins
PTGS2 (COX-2) Recombinant Human Protein
Lyophilized
68.5kd
5μg
Full Length, Recombinant
Prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase and cyclooxygenase)
Assay Standard
Insect
His Tag
Videos