Learn More
DWK Life Sciences DURAN™ Laboratory Thread Adapters
Brand:  DWK Life Sciences 1168446
Code : A0
Additional Details : Weight : 0.04900kg
 View more versions of this product
Product Code. 10326553
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Adapters are available for GL 40, GL 38 and GL 32 screw threads.
Specifications
Specifications
1Each
Thread Adapter
PTFE
Thread Adapter
Handling aggressive, gas generating media
GL38/GL45
Videos