missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ PTFE Spinbar
OHAUS™ PTFE Stirbar for stirring volumes in vessels such as vials and tubes.
£9.80 - £76.70
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
15792559
|
OHAUS™
30400147 |
2.5 cm |
£9.80
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
15702569
|
OHAUS™
30400148 |
3.8 cm |
£11.20
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
15712569
|
OHAUS™
30400149 |
7 cm |
£40.30
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
15722569
|
OHAUS™
30400150 |
10 cm |
£76.70
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
- Constructed of PTFE
- 10 cm spinbar includes a pivot ring
Specifications
White | |
PTFE |