Learn More
JG Finneran Associates™ PTFE Seals
Use these inserts with dram sample vials. JG Finneran™ dram sample vials are made of Type I borosilicate glass, providing exceptional resistance to heat shock and chemical leaching.
Brand: JG Finneran Associates™ 601010-08
Code : 17
Additional Details : Weight : 0.88360kg
Description
- PTFE seals
- For use with dram sample vials
- Available in a variety of sizes
Specifications
PTFE | |
White | |
1000 Pk. | |
8 mm |