Learn More
BRAND™ PTFE Seal
For use with the Transferpettor™ line, specially designed to handle viscous, foaming liquids and high vapor pressure.
Brand: BRAND™ 701924
Code : 03
Additional Details : Weight : 0.01000kg
Description
- With DE-M marking
- Provided with mounting block
- Provided in packs of 3
Specifications
Conformity certified | |
100 and 200 μL | |
Mounting block |
PTFE | |
For Transferpettor™ 100, 200µL capacity | |
3/Pk. |