missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cowie Technology™ PTFE Pointed Tweezers
Cowie Technology™ PTFE Pointed Tweezers are inert to virtually all chemical reagents and completely non-contaminating.
Brand: Cowie Technology™ 027.201
Code : 41
Additional Details : Weight : 0.08400kg
Description
- Chemically inert
- Non-contaminating
- Usable to 280°C
- Manufactured from Virgin PTFE materials with FDA and USP Class VI approvals
Specifications
Pointed | |
PTFE | |
PTFE Forceps / Tweezers | |
12 mm | |
25 mm | |
2.5 mm | |
25 mm |
1 Each | |
200 mm | |
Smooth | |
12 | |
11 mm | |
200 x 25 mm | |
Rigid |