Learn More
Fisherbrand™ PTFE Platinum Resistance Probe with 1M Cable
Inert to virtually all chemical reagents. Fisherbrand™ PTFE Thermocouple Type K Probe combine the outstanding properties of PTFE with the accuracy and reliability of platinum resistance and thermocouple thermometry.
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 0.10000kg
Description
- Non-contaminating
- Impervious to chemical diffusion at high pressure
- Wide operating temperature range, -180 to +300°C
- Outstanding electrical insulation properties
- Individually tested
Specifications
Probe, PTFE Platinum Resistance | |
PTFE with Stainless Steel Core | |
Measuring temperature | |
50%, 6 to 14 sec. | |
300 mm | |
100 g |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PRT100 Class A. TCR = 3850 ppm/degree celsius | |
PTFE Probe Holders and Compression Fittings | |
-180°C to +280°C | |
8 mm |