missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ PTFE One-Sided Gasket

Use this washer as a gasket for BOLA Screw Caps with Aperture (Cat. No. H 983-../ H 984-./ H 995-…) Bohlender™ BOLA™ PTFE One-Sided Gaskets are also suitable for Duran Group GL caps (formerly Schott AG).

Brand:  Bohlender™ 292382602

Code : A0

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 11777118

  • £158.00 / Pack of 10


Estimated Shipment
03-07-2018
Add to basket

Description

Description

  • Silicone ring with PTFE washer
  • After assembly, the product is only exposed to PTFE
  • Universal chemical resistance
  • Temperature range: -60 to +230°C
  • Packing unit: Ten pieces, differing ordering quantities are rounded up to factor 10
Specifications

Specifications

For BOLA Screw Caps and also Suitable for GL Caps
PTFE
42 I.D. x 26 O.D. x 9 Hmm
9mm
100Pack
One-sided Gaskets
Videos
Documentation