Learn More
Fisherbrand™ PTFE Micro Magnetic Stir Bar
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
Brand:  Fisherbrand™
Code : 41
Additional Details : Weight : 0.00270kg
 View more versions of this product
Product Code. 11868882
  • £12.40 / Pack of 10
In Stock
to see stock.
Add to basket
Description
Description
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
  • Supplied in reusable snap cap vials.
  • For stirring in test tubes, small size beakers, or other containers.
  • Autoclavable.
Specifications
Specifications
3 mm
PTFE encapsulated Alnico V magnet core
White
Containers & Vessels / Stirrer Hot Plates
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Cylinder
PTFE
PTFE Stirrer Bars Micro
10/Pk.
Yes
3 mm