missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Micro Magnetic Stir Bar

Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 0.00085kg

 View more versions of this product

Product Code. 10652401

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Supplied in reusable snap cap vials.
  • For stirring in test tubes, small size beakers, or other containers.
  • Autoclavable.
Specifications

Specifications

Yes
PTFE
PTFE encapsulated Alnico V magnet core
10/Pk.
3mm
Cylindrical
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
White
Containers & Vessels / Stirrer Hot Plates
PTFE Stirrer Bars Micro
10mm
Videos