Learn More
Fisherbrand™ PTFE Conical Joint Clamps
Use as a safe solution to clamping glass joints. The PTFE conical joint clamps include an encapsulated steel spring.
Brand:  Fisherbrand™
Code : 41
Additional Details : Weight : 0.02200kg
 View more versions of this product
Product Code. 10138231
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
PTFE joint clamps are chemically inert and chemically resistant. Temperature resistant, maintaining strength up to 280°C (480°F). Manufactured from pure PTFE, with totally encapsulated steel springs. They can be adjusted by hand and fit standard glass joints.
Specifications
Specifications
PTFE Encapsulated Steel Spring
White
PTFE Joint Clamp
Conical
Standard glass joints, size 29
2Pack
Videos