Learn More
Fisherbrand™ PTFE Joint Adapter
Creates high vacuum with sealing rings, negating the need for grease. Cowie Technology™ PTFE Joint Adapter are autoclavable and reusable.
Brand:  Fisherbrand™
UNSPSC : 41121815
Code : 33
Additional Details : Weight : 0.06400kg
 View more versions of this product
Product Code. 11543342
  • £35.20 / Pack of 2
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Description
Made from PTFE with a substantial shoulder for ease of handling and an overall robust construction for repeated use. Rigid sleeves and adapters are inert, usable to 280°C (approx.) and will withstand high vacuum. High vacuum is obtained using transverse trapezoidal sealing rings on the contacting surfaces. For grease free, high vacuum seals using standard ground glass joints.
Description & Specifications
Specifications
1Each
PTFE Joint Adapters
Documentation