Learn More
Fisherbrand™ PTFE Joint Adapter
Creates high vacuum with sealing rings, negating the need for grease. Cowie Technology™ PTFE Joint Adapter are autoclavable and reusable.
Brand:  Fisherbrand™
Code : 33
Additional Details : Weight : 0.13600kg
 View more versions of this product
Product Code. 11513342
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Made from PTFE with a substantial shoulder for ease of handling and an overall robust construction for repeated use. Rigid sleeves and adapters are inert, usable to 280°C (approx.) and will withstand high vacuum. High vacuum is obtained using transverse trapezoidal sealing rings on the contacting surfaces. For grease free, high vacuum seals using standard ground glass joints.
Specifications
Specifications
1Each
PTFE Joint Adapters
Videos
Documentation