missing translation for 'onlineSavingsMsg'
Learn More

Brand™ PTFE Ground Joint Sleeves

Brand:  Brand™ 51430

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12085332

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Joint clips
Ground Joint Sleeves
–200 to 260°C
NS 40/38
PTFE
10Pack
0.05mm
Ground Joint Sleeves
Videos