Learn More
Kartell™ Lining for Glass Tapers
Constructed of PTFE for high temperature and chemical resistance. Kartell™ Lining for Glass Tapers are specially designed to prevent machined glass joints from binding.
Brand:  Kartell™ 0104100
Code : NEW
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10717741
  • £34.25 / Pack of 10
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
Description
Description
  • Designed to prevent machined glass joints from binding
  • Conforms to DIN 12242
  • Suitable for foodstuff
Specifications
Specifications
Ground Joint Sleeve Lining
PTFE
Glass Joint
10/Pk.
DIN 12242
26 mm
Prevent glass joints from binding
29/32 Cone Size