missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Welch Ilmvac™ PTFE Filter
Brand: Welch Ilmvac™ 828840-20
Code : NEW
Additional Details : Weight : 0.10000kg
Specifications
55 mm | |
PTFE | |
1 μm |
Biovac 060 and Biovac 106 | |
10 Pack | |
Filter |