Learn More
Bohlender™ PTFE Point Tip Tweezers
Use these tweezers for a variety of applications. BOLA PTFE Tweezers feature universal chemical resistance.
Brand: Bohlender™ H909-02
Additional Details : Weight : 0.10000kg
Description
- Precast tweezers
- PTFE construction
- With pointed ends
- Universal chemical resistance
Specifications
Pointed | |
Precast tweezer | |
100 mm |
Each | |
PTFE | |
Tweezers |