missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Blade for Plain Shafts

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.01880kg

 View more versions of this product

Product Code. 10331352

  • £3.50 / Each


In Stock 5
Add to basket

Description

Description

  • Made of PTFE
  • Totally inert and non-scratch
  • Available in square end and plain end format
Specifications

Specifications

PTFE Blades for PTFE Shafts & Complete PTFE Blade with PTFE Shaft
Stirring in Narrow Neck Vessel
PTFE
3mm
15g
1Each
PTFE Shaft Stirrer, Plain Shaft for Blades
-200 to +280°C
90mm
28mm
Videos