missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Blade for Plain Shafts

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 10564552

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Made of PTFE
  • Totally inert and non-scratch
  • Available in square end and plain end format
Specifications

Specifications

Blades for Plain Shafts
Stirring in Narrow Neck Vessel
PTFE
3mm
4g
1Each
PTFE Shaft Stirrer, Plain Shaft for Blades
-200 to +280°C
52mm
14mm
Videos