Fisherbrand™ PTFE Beaker with Spout

Includes a machined base for good heat transfer. The PTFE beaker includes a molded-in spout for easy pouring.

Overview
Brand: Fisherbrand™

UNSPSC: 41121803

Code: 41

Additional Details:
Additional Details: Weight: 0.00600kg



Product Code. 10733271

Quantity Price
1 £ 6.27 / Each
Estimated Shipment
24-03-2017
Add to basket


Description and Specification

Specification

ISO/DIN 7056 No
Product Type Beaker
Material PTFE (Polytetrafluoroethylene)
Diameter (Metric)Top 20mm
Capacity (Metric) 5mL
Disposable No
Temperature (Metric) Max. Operating 280°C
Autoclavable Yes
Color White
Diameter (Metric) Outer 20mm
Graduated No
Height (Metric) 26mm
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)

Non-contaminating and inert to highly corrosive reagents.
  • Non-wettable, lightweight, and heat resistant up to 280°C
  • Machined base for good heat transfer
  • Easy to clean
  • Molded-in spout for easy pouring
  • Isostatically molded from pure PTFE

Documentation