Learn More
Fisherbrand™ PTFE Beaker Cover
Catalog Number 10702931
Click to view available options
Diameter: 40mm
Rest on top of beaker to prevent contamination. The PTFE Beaker Cover is available in a wide range of sizes.
  • Inert, chemically resistant
  • Smooth internal finish
  • Exceptional thermal resistance: can be used down to -200°C and up to 280°C
  • Isostatically molded from pure PTFE