missing translation for 'onlineSavingsMsg'
Learn More

Lufft Mess und Regeltechnik™ PT100 Immersion Probe

Brand:  Lufft Mess und Regeltechnik™ 3120.520

UNSPSC : 41110000

Code : NEW

Additional Details : Weight : 0.12000kg

 View more versions of this product

Product Code. 15309071

  • POA / Each


Estimated Shipment
04-08-2017
Add to basket

Specifications Description & Specifications

Specifications

Hand-held measuring device XP100
measuring range: -40 to 400°C, storage temperature: -40 to 60°C, operating temperature up to 80°C
Suitable for measurements in gaseous media, liquids and granular material, such as sand
150mm