missing translation for 'onlineSavingsMsg'
Learn More

Retsch™ PT 200 Model Rotating Tube Dividers

Brand:  Retsch™ 40.412.0002

Code : NEW

Additional Details : Weight : 47.00000kg

 View more versions of this product

Product Code. 11994192

  • £9146.67 / Each


Estimated Shipment
27-02-2018
Add to basket

Specifications

Specifications

551 x 520 x 1050mm
Field of application agriculture, biology, chemistry/Plastics, construction materials, engineering/electronics, environment/recycling, food, geology/metallurgy, glass/ceramics, medicine/pharmaceuticals
551mm
1-phase Power Connection
520mm
Rotating Tube Divider
220/240V, 50Hz
1050mm
PT 200
~46kg
50Hz
220/240V
Videos