missing translation for 'onlineSavingsMsg'
Learn More

Psora 4, Tocris Bioscience™

Potent KV1.3 channel blocker

Brand:  RD Systems (Bio-Techne) 4367/50

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 50mg

 View more versions of this product

Product Code. 13454634

  • £787.92 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent KV1.3 channel blocker (EC50 = 3 nM). Preferentially binds the C-type inactivated state of the channel, similar to the activity of CP 339818 (Cat. No. 1399). Displays 17- to 70-fold selectivity over closely-related KV1 channels (KV1, KV2, KV4 and KV7); exhibits activity at KV1.5 (IC50 = 7.7 nM). Suppresses proliferation of human and rat myelin-specific effector memory T cells (EC50 values are 25 and 60 nM respectively).
Specifications

Chemical Identifiers

724709-68-6
334.371
psora-4, psora 4, 5-4-phenylbutoxy psoralen, 4-4-phenylbutoxy furo 3,2-g chromen-7-one, 4-4-phenylbutoxy-7h-furo 3,2-g chromen-7-one, 4-4-phenylbutoxy-7h-furo 3,2-g 1 benzopyran-7-one, lopac-p-9872, d01nck, d0h1ei, psora hplc
4-(4-phenylbutoxy)furo[3,2-g]chromen-7-one
C21H18O4
JJAWGNIQEOFURP-UHFFFAOYSA-N
6603977
C1=CC=C(C=C1)CCCCOC2=C3C=CC(=O)OC3=CC4=C2C=CO4

Specifications

Psora 4
724709-68-6
psora-4, psora 4, 5-4-phenylbutoxy psoralen, 4-4-phenylbutoxy furo 3,2-g chromen-7-one, 4-4-phenylbutoxy-7h-furo 3,2-g chromen-7-one, 4-4-phenylbutoxy-7h-furo 3,2-g 1 benzopyran-7-one, lopac-p-9872, d01nck, d0h1ei, psora hplc
C1=CC=C(C=C1)CCCCOC2=C3C=CC(=O)OC3=CC4=C2C=CO4
334.371
334.37
50mg
C21H18O4
JJAWGNIQEOFURP-UHFFFAOYSA-N
4-(4-phenylbutoxy)furo[3,2-g]chromen-7-one
6603977
>99%
Videos
Documents