missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSGL-1/CD162 Mouse anti-Human, Clone: CHO131, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
POA
Specifications
Antigen | PSGL-1/CD162 |
---|---|
Clone | CHO131 |
Immunogen | CHO Chinese hamster ovary cell line transfected with human PSGL-1/CD162 |
Gene ID (Entrez) | 6404 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Human samples. It is validated for Flow Cytometry, Adhesion Blockade, CyTOF-ready.Specifications
PSGL-1/CD162 | |
CHO Chinese hamster ovary cell line transfected with human PSGL-1/CD162 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human PSGL-1/CD162. Recognizes sLe x -bearing core 2 O-glycan stuctures. It does not recognize sLe x on an extended core 1 O-glycan. The sLe x -bearing, core 2 O-glycan structure decorates the P-Selectin ligand PSGL-1, and the presence of this glycan structure is required for high affinity P-Selectin binding (1). This antibody stains human and canine leukocytes but does not recognize monkey, mouse, rabbit, porcine, feline or bovine leukocytes. | |
Unconjugated | |
Flow Cytometry, Adhesion Blockade, CyTOF | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
CHO131 | |
6404 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgM | |
Affinity Purified | |
RUO |
For Research Use Only