Learn More
PSF™ Leather 764SM Model Boots
Brand:  PSF Progressive Safety 764SM-10
Code : 91
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12724486
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
EN ISO 20345:2011
Oils, fats, petrol, alkalis and heat, water
1kg
Leather
Boots
Nitrile Rubber Outsole, Cushioned Polyurethane Midsole
Black
10
Videos