0 PSF™ Contractor Safety Shoes


Overview
Brand: 0

Manufacturer Part Number: 806SM-13

1 PAIR Safety shoes Contractor 806SM with steel

UNSPSC: 46181605

Code: 91

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 12817583

Quantity Price
1 £ 48.0 / 1 pair
Estimated Shipment
30-03-2017
Add to basket


Description and Specification

Specification

Toe Material Steel
Size 13
Product Type Contractor Safety Shoes
Resistant Heat, Oils and General Chemicals
Style Contractor Safety
Sole Features Dual Density
Color Black
Material Leather Upper
For Use With (Application) Industry, Maintenance, College, Mechanics
Sole Material Polyurethane
Certifications/Compliance S1 Rated