missing translation for 'onlineSavingsMsg'
Learn More

PSB 11 hydrochloride, Tocris Bioscience™

Potent, selective hA3 receptor antagonist/inverse agonist

Brand:  Tocris (Bio-Techne) 201210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13440404

  • £178.20 / 10mg

Estimated Shipment
02-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective antagonist for the human adenosine A3 receptor, with low affinity for the rat A3 receptor (Ki values are 2.3 and > 10000 nM respectively). Displays > 1000-fold selectivity over human A1 and A2A receptors (Ki values are 4.1 and 3.3 μM respectively) and > 180-fold selectivity over rat A1, rat A2A and mouse A2B receptors. Acts as an inverse agonist in the [35S]GTPγS binding assay in hA3-CHO cells (IC50 = 36 nM).
Specifications

Chemical Identifiers

453591-58-7
331.804
psb 11 hydrochloride, 8r-8-ethyl-1,4,7,8-tetrahydro-4-5h-imidazo 2,1-i purin-5-one hydrochloride, 8r-8-ethyl-4-methyl-2-phenyl-1,4,7,8-tetrahydro-5h-imidazo 2,1-i purin-5-one-hydrogen chloride 1/1
(8R)-8-ethyl-4-methyl-2-phenyl-7,8-dihydro-1H-imidazo[2,1-f]purin-5-one;hydrochloride
C16H18ClN5O
JAUSHKIWPLHBCN-RFVHGSKJSA-N
16759152
CCC1CN2C(=N1)C3=C(N=C(N3)C4=CC=CC=C4)N(C2=O)C.Cl

Specifications

PSB 11 hydrochloride
453591-58-7
psb 11 hydrochloride, 8r-8-ethyl-1,4,7,8-tetrahydro-4-5h-imidazo 2,1-i purin-5-one hydrochloride, 8r-8-ethyl-4-methyl-2-phenyl-1,4,7,8-tetrahydro-5h-imidazo 2,1-i purin-5-one-hydrogen chloride 1/1
CCC1CN2C(=N1)C3=C(N=C(N3)C4=CC=CC=C4)N(C2=O)C.Cl
331.804
331.8
10mg
C16H18ClN5O
JAUSHKIWPLHBCN-RFVHGSKJSA-N
(8R)-8-ethyl-4-methyl-2-phenyl-7,8-dihydro-1H-imidazo[2,1-f]purin-5-one;hydrochloride
16759152
>98%
Videos