missing translation for 'onlineSavingsMsg'
Learn More

PSB 0739, Tocris Bioscience™

Highly potent P2Y12 antagonist

Brand:  RD Systems (Bio-Techne) 3983/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13453924

  • £196.98 / 10mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Highly potent P2Y12 receptor antagonist (Ki = 24.9 nM). Unlike clopidogrel (Cat. No. 2490), does not require bioactivation.
Specifications

Chemical Identifiers

1052087-90-7
611.551
1-Amino-9, 10-dihydro-9, 10-dioxo-4-[[4-(phenylamino)-3-sulfophenyl]amino]-2-anthracenesulfonic acid sodium salt
1-amino-4-(4-anilino-3-sulfoanilino)-9,10-dioxoanthracene-2-sulfonic acid;sodium
C26H19N3Na2O8S2
CVHGFNQCVCSUDU-UHFFFAOYSA-N
87467252
C1=CC=C(C=C1)NC2=C(C=C(C=C2)NC3=CC(=C(C4=C3C(=O)C5=CC=CC=C5C4=O)N)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]

Specifications

10mg
1052087-90-7
1-Amino-9, 10-dihydro-9, 10-dioxo-4-[[4-(phenylamino)-3-sulfophenyl]amino]-2-anthracenesulfonic acid sodium salt
C1=CC=C(C=C1)NC2=C(C=C(C=C2)NC3=CC(=C(C4=C3C(=O)C5=CC=CC=C5C4=O)N)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
611.551
609.54
PSB 0739
C26H19N3Na2O8S2
CVHGFNQCVCSUDU-UHFFFAOYSA-N
1-amino-4-(4-anilino-3-sulfoanilino)-9,10-dioxoanthracene-2-sulfonic acid;sodium
87467252
>96%
Videos
Documents