missing translation for 'onlineSavingsMsg'
Learn More

Proxyfan oxalate, Tocris Bioscience™

High affinity H3 ligand

Brand:  Tocris (Bio-Techne) 247750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13451294

  • £501.27 / 50mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

High affinity histamine H3 receptor ligand (pKi = 8.62) that acts as a protean agonist at recombinant and native receptors. In functional studies, activity ranges from full agonist to inverse agonist depending on system used. Displays partial agonist effects on cAMP inhibition and MAPK activity, neutral antagonist activity on histamine release and partial inverse agonism of [3H]-arachidonic acid release.
Specifications

Chemical Identifiers

177708-09-7
C13H16N2O·C2H2O4
but-2-enedioic acid-5-3-benzyloxy propyl-1h-imidazole 1/1

Specifications

50mg
177708-09-7
but-2-enedioic acid-5-3-benzyloxy propyl-1h-imidazole 1/1
>98%
Proxyfan oxalate
C13H16N2O·C2H2O4
306.32
Videos
Documents