Learn More
Macherey Nagel Bioanalysis™ Protino™ Gst/4B Column
Best choice for cost effective GST-tag protein purification in ready to use 5 mL FPCL columns
Brand: Macherey Nagel Bioanalysis™ 745515.1
Code : 86
Additional Details : Weight : 0.00500kg
Description
5mL Macherey-NagelSpecifications
4°C to 8°C | |
90 μm | |
Purification of GST-tag proteins | |
FPLC Columns |
Protein solution | |
1 Piece | |
Protein |
Safety and Handling
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Supplemental information
- MIXTURE LIST-Contains : ethanol (diluted < 20 %) (denatured with1%IPA/1%MEK, acc.2016/1867/EU)