Learn More
Thermo Scientific™ Proteinase K, Ready-to-Use Soln.
A stable and highly reactive serine protease; used for protein and nucleic acid isolation
Brand: Thermo Scientific™ J63710.AC
Code : NEW
Additional Details : CAS Number : 39450-01-6 Weight : 0.00025kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
39450-01-6 | |
-30°C to -10°C | |
A stable and highly reactive serine protease; used for protein and nucleic acid isolation | |
25 mL |
Clear, Colorless | |
28.9 kd | |
DNA Extraction Reagent | |
Liquid,28.9 kDa |